Order Entry
Export
Orders LinkContactUsLinkComponent
Triethyl citrate

Triethyl citrate

Glass bottle, 1 L
Catalog # SIAL27500-1L-F
Supplier:   Merck
Triethyl citrate
Catalog # SIAL27500-1L-F
Supplier:  Merck
CAS Number:  77-93-0
Triethyl citrate

Glass bottle, 1 L

CAS Number:  77-93-0
Price
209,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
  • CAS Number:77-93-0
  • Formula:C₁₂H₂₀O₇
  • MDL Number:MFCD00009201
  • Molecular Weight:276.29 g/mol
  • Boiling Pt:294 °C (1013 hPa)
  • Melting Pt:–55 °C
  • Density:1.135 g/cm³ (25 °C)
  • Flash Pt:150 °C
  • EINECS:201-070-7
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications