Immersion oil for microscopy, Sigma-Aldrich®
Immersion oil, 250 mL
Supplier: SIGMA ALDRICH MICROSCOPY
Immersion oil for microscopy, Sigma-Aldrich®
Catalog # SIAM56822-250ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Jump to:
Product Details & Documents
Immersion oil is used for high resolution (1000X) light microscopy work under oil immersion objective lens.
- Suitable for Hematology
- Suitable for Histology
Documents
Specifications
Specifications
- Pk:250 mL
- Description:Immersion oil
- Viscosity:100-120 mPa.s (20 °C)
- Cat. No.:SIAM56822-250ML
- M-Clarity quality level:MQ100
- Refractive index:n20/D 1.516
- MDL No.:MFCD00131369
- Density:1.025 g/mL (20 °C)



