0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Copper
Copper, Sigma-Aldrich®
Copper, Sigma-Aldrich®
Glass bottle for solids, 1 kg
SIAL61139-1KG
:
Merck
Copper, Sigma-Aldrich®
SIAL61139-1KG
:
Merck
:
7440-50-8
Copper, Sigma-Aldrich®
Glass bottle for solids, 1 kg
:
7440-50-8
264,00 €
1 * 1 kg
-
+
:
7440-50-8
:
Cu
:
MFCD00010965
:
63.55 g/mol
:
13,02545
:
2595 °C (1013 hPa)
:
1083 °C
:
8.94 g/cm³ (20 °C)
:
Ambient
:
231-159-6
Pack type:
Glass bottle for solids
Pk:
1 kg