Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(-)-Fructose, Sigma-Aldrich®

Glass bottle for solids, 1 kg
Catalog # SIAL47739-1KG-F
Supplier:   Merck
D-(-)-Fructose, Sigma-Aldrich®
Catalog # SIAL47739-1KG-F
Supplier:  Merck
CAS Number:  57-48-7
D-(-)-Fructose, Sigma-Aldrich®
D-(-)-Fructose, Sigma-Aldrich®

Glass bottle for solids, 1 kg

CAS Number:  57-48-7
Price
331,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications