Orders LinkContactUsLinkComponent
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extractor, 24-400 screw thread on top and 20-400 screw thread on bottom, — ml
Catalog # WHEA415502
Supplier:   DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Catalog # WHEA415502
Supplier:  DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extractor, 24-400 screw thread on top and 20-400 screw thread on bottom, — ml
Price
384,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifications
  • Description:
    Soxhlet extractor, 24-400 screw thread on top and 20-400 screw thread on bottom
  • Seq:
    03
  • Flask capacity:
    — ml