Orders LinkContactUsLinkComponent
Disposable earplugs, E-A-RSoft™ FX
Disposable earplugs, E-A-RSoft™ FX
Earplug, with orange cord, Light green
Catalog # MMMAES01021
Supplier:   3M
Disposable earplugs, E-A-RSoft™ FX
Catalog # MMMAES01021
Supplier:  3M
Disposable earplugs, E-A-RSoft™ FX
Disposable earplugs, E-A-RSoft™ FX
Earplug, with orange cord, Light green
Price
124,00 €
Log in for availability
1 * 200 PAIR
Product Details & Documents
This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industry. Custom polyurethane foam formulation provides slow recovery and extreme softness for optimum comfort.
  • Flange shape serves as an effective insertion device when rolled down
  • Easy to fit into the ear canal
  • Available with or without cord
  • Compatible with other PPE
SNR: 39 dB
Specifications
  • Colour:
    Light green
  • Description:
    Earplug, with orange cord