Disposable earplugs, E-A-RSoft™ FX
Earplug, without cord, Light green
Catalog # 111-0835
Supplier: 3M
Jump to:
Product Details & Documents
This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industry. Custom polyurethane foam formulation provides slow recovery and extreme softness for optimum comfort.
- Flange shape serves as an effective insertion device when rolled down
- Easy to fit into the ear canal
- Available with or without cord
- Compatible with other PPE
SNR: 39 dB
Specifications
- Colour:Light green
- Description:Earplug, without cord