Orders LinkContactUsLinkComponent
Sampling valves
Sampling valves
Valve outlet cap with needle
Catalog # MILXMXPESP5CN
Supplier:   Merck
Sampling valves
Catalog # MILXMXPESP5CN
Supplier:  Merck
Sampling valves
Sampling valves
Valve outlet cap with needle
Price
55,70 €
Log in for availability
1 * 5 items
Product Details & Documents
The sampling valves are used for sterile sampling of water or other final products in the pharmaceutical industry.
  • Collect samples mid-stream and filter through a MicropreSure unit, or sample into sterile sampling bags or a graduated cylinder
  • Samples are kept in sanitary condition for immediate sampling with alcohol or hydrogen peroxide injections
Materials: stem and body: 316L stainless steel or ECTFE body in ECTFE-type units, valve cap: ABS or PP, O-ring and plug seal: silicone or fluoroelastomer.
Certifications: Meets the FDA indirect food requirements cited in code of federal regulations CFR 21, part 177-182.
Specifications
  • Description:
    Valve outlet cap with needle
  • Seq:
    11