Biotin kits, HOOK™
Iodoacetyl-LC-Biotin kit, 1 KIT
Catalog # BS-12
Supplier: G-Biosciences
Jump to:
Product Details & Documents
HOOK™ Biotin Reagent that conjugates to proteins through amines, sulfhydryls, carboxyls or carbohydrates. The amine and sulfhydryl coupling HOOK™ Biotin Reagents couple directly to the protein through their reactive groups, however the carboxyl coupling HOOK™ Biotin Reagents require a carbodiimide crosslinker and the carbohydrate coupling HOOK™ Biotin Reagents require carbohydrate oxidation before coupling.
- Optimizer Buffer™ for improved coupling efficiency
- SpinOUT™ gel filtration columns for rapid (<10 minutes) purification
- Biotin assay reagents to determine level of biotin incorporation
- Labels 1 to 10 mg protein/reaction
- Suitable for 10 coupling reactions
The kits include EDC as the carbodiimide crosslinker in the carboxyl coupling kits and sodium meta-periodate for carbohydrate oxidation in the carbohydrate coupling kits. Kit also contains a specific Optimizer Buffer™ that provides the optimal reaction conditions for each HOOK™ Biotin Reagent.
Documents
Specifications
Specifications
- Pk:1 KIT
- Description:Iodoacetyl-LC-Biotin kit
Specifications
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-V, 5X
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-III, 5X
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-IV, 5X
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-VI, 5X
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-II,5X
Product Information
- Catalog #PkAvailabilityPrice
- Optimizer Buffer™-I, 5X