Detergent removal system, DetergentOUT™ Tween®
DetergentOUT™ Tween®, Micro, 10
Supplier: G-Biosciences
Jump to:
Product Details & Documents
DetergentOUT™ Tween® is a spin column format detergent removal resin for polysorbate or Tween® detergents or surfactants.
DetergentOUT™ Tween® specifically removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
Documents
Specifications
Specifications
- Pk:10
- Description:DetergentOUT™ Tween®, Micro