Orders LinkContactUsLinkComponent
Disposable cannulas
Disposable cannulas, G22, Black, 50 mm, +, 0,7×50 mm
Catalog # EHRS15884650
Supplier:   EHRHARDT SOEHNE
Disposable cannulas
Catalog # EHRS15884650
Supplier:  EHRHARDT SOEHNE
Disposable cannulas
Disposable cannulas
Disposable cannulas, G22, Black, 50 mm, +, 0,7×50 mm
Price
32,40 €
Log in for availability
1 * 100 items
Product Details & Documents
Disposable cannulas.
  • Pyrogen-free, not toxic
  • With lancet cut
  • EO-sterilisation
Specifications
  • Ø×L:
    0,7×50 mm
  • Gauge:
    G22
  • Sterile:
    +
  • Colour:
    Black
  • Description:
    Disposable cannulas
  • Length:
    50 mm
  • Seq:
    04