Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dichloropurine

2,6-Dichloropurine

Bottle, 25 g
Catalog # APOSOR11256-25G
Irritant
Supplier:   Apollo Scientific
2,6-Dichloropurine
Catalog # APOSOR11256-25G
Supplier:  Apollo Scientific
CAS Number:  5451-40-1
2,6-Dichloropurine

Bottle, 25 g

CAS Number:  5451-40-1
Price
31,90 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2,6-Dichloro-9H-purine
  • CAS Number:5451-40-1
  • Formula:C₅H₂Cl₂N₄
  • MDL Number:MFCD00077725
  • Molecular Weight:189 g/mol
  • Melting Pt:184…186 °C
  • Storage Temperature:Ambient
  • EINECS:226-681-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications