Order Entry
Export
Orders LinkContactUsLinkComponent
Tungsten trioxide 99+%

Tungsten trioxide 99+%

Plastic bottle, 500 g
Catalog # ACRO214215000
Irritant
Tungsten trioxide 99+%
Catalog # ACRO214215000
CAS Number:  1314-35-8
Tungsten trioxide 99+%
Tungsten trioxide 99+%

Plastic bottle, 500 g

CAS Number:  1314-35-8
Price
274,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Tungsten (VI) oxide
  • CAS Number:1314-35-8
  • Formula:O₃W
  • MDL Number:MFCD00011466
  • Molecular Weight:231.84 g/mol
  • Merck Index:12,09947
  • Melting Pt:1470…1475 °C
  • Density:7,16 g/cm³ (20 °C)
  •  :Density: 7.16 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-231-4

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to yellow-green Powder
  • Assay:
    ≥99.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications