Order Entry
Export
Orders LinkContactUsLinkComponent
Jars with screw cap

Jars with screw cap

with polypropylene screw cap, 150 ml
Catalog # 216-2046
Jars with screw cap
Catalog # 216-2046
Supplier:  
Jars with screw cap
Jars with screw cap

with polypropylene screw cap, 150 ml

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Container Con.Specimen 120 ml PP with white screw cap 1 * 300 items Catalog #216-0863

Product 216-0863 is similar to 216-2046 but manufacturer and specifications may vary.

Product Details & Documents

Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.

  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Type:
    with polypropylene screw cap
  • Capacity:
    150 ml
  • Seq:
    09