Orders LinkContactUsLinkComponent
Wound cleansers
Wound cleansers
Salvequick cleaning wipes refill box, incl. 20 pcs sterile wound cleansers (110×120 mm)
Catalog # 118-0250
Supplier:   ORKLA CARE
Wound cleansers
Catalog # 118-0250
Supplier:  ORKLA CARE
Wound cleansers
Wound cleansers
Salvequick cleaning wipes refill box, incl. 20 pcs sterile wound cleansers (110×120 mm)
Price
9,10 €
Log in for availability
1 * 20 items
Product Details & Documents
Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (0,9% NaCl), are intended for the mechanical cleansing of minor wounds when clean tap water is not available.
  • Tissues are packed in sealed sachets, which prevent leakage and guarantee sterility
  • Savett sachets are packed in a refill box
  • Salvequick cleansers fit into first-aid-stations
Documents
Specifications
  • Description:
    Salvequick cleaning wipes refill box, incl. 20 pcs sterile wound cleansers (110×120 mm)