Order Entry
Export
Orders LinkContactUsLinkComponent
 

Retort rings

Ring without bosshead, 260 g, 160 mm
Catalog # JUCHVL04930
Retort rings
Catalog # JUCHVL04930
Retort rings

Ring without bosshead, 260 g, 160 mm

Price
19,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Steel, epoxy coated.

  • Available with or without bosshead
  • Bosshead is made of malleable iron
Specifications
  • Ext.Ø:
    160 mm
  • Description:
    Ring without bosshead, 260 g
  • Seq:
    05