Double bossheads, cross shape
Double bosshead, cross shape, with M8 brass thumb bolt, Iron, 16 mm
Catalog # JUCHVL04850
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Bossheads made from either epoxy coated malleable iron or from stainless steel.
Specifications
- Description:Double bosshead, cross shape, with M8 brass thumb bolt
- Material:Iron
- For rod Ø:16 mm