Orders LinkContactUsLinkComponent
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da., 1 g, 1
Catalog # VARIPL2024-2001
Supplier:   VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Catalog # VARIPL2024-2001
Supplier:  VARIAN
CAS Number:  
Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da., 1 g, 1
CAS Number:  
Price
223,01 €
Log in for availability
1 * 1 items
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
  • Many solvent options increase applicability
  • Stringent quality control improves performance
  • Proprietary manufacturing methods ensure consistent supply
Specifications
Specifications
  • Pk:
    1
  • Description:
    Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,09, nominal Mp 1,500,000 Da.
  • Pk Info:
    1 g
Specifications