ContactUsLinkComponent
Tube roller RM10W-30V, 10 removable rollers synchronous rocking and rotating mixer, variable speed 1 - 30 rpm, roll length: 330 mm, diameter 32 mm, average distance from roll to roll: 36.5 mm, tilt angle: 3,138 1 * 1 items
Catalog # 444-0868
Supplier:  CAT
undefined
Tube roller RM10W-30V, 10 removable rollers synchronous rocking and rotating mixer, variable speed 1 - 30 rpm, roll length: 330 mm, diameter 32 mm, average distance from roll to roll: 36.5 mm, tilt angle: 3,138 1 * 1 items
Catalog # 444-0868
Supplier:  CAT