Automation tips for Tecan®, Axygen®
EV-205-WB-R (wide bore), 200 µl, -, -, Transparent, MCA96 / HV96, 50 racks, 96 each
Catalog # 736-0523
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
PP, with or without filter, sterile or non sterile.
- Black tips are conductive for liquid level-sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
: Packaged in plastic blister packs.
: * Economy packed in cardboard wrapper.
Specifications
- Packed:50 racks, 96 each
- Sterile:-
- Volume:200 µl
- Seq:34
- Tip style:EV-205-WB-R (wide bore)
- Tip colour:Transparent
- For use with:MCA96 / HV96
- Filter:-