Orders LinkContactUsLinkComponent
VWR® LA Classic, Analytical Balances, LCD
LA 124 *, 120 g, 0,1 mg, Ø 80 mm, 0,3 mg
  611-2258
VWR® LA Classic, Analytical Balances, LCD
  611-2258
VWR® LA Classic, Analytical Balances, LCD
VWR® LA Classic, Analytical Balances, LCD
LA 124 *, 120 g, 0,1 mg, Ø 80 mm, 0,3 mg
1 480,00 €
1 * 1 items
An economical range of reliable, high performance analytical balances. All models have a stainless steel top pan.
  • Check weighing (alarm function), weight totalising, density determinations of samples and piece counting functions
  • Option of external or internal calibration, RS232 included as standard
  • Weighing chamber L×W×H: 162×171×225 mm
User can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight.
 : EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012.
 : Optional Tablet System (611-8683) is suitable for use with models denoted by *. Tablet Systems are not currently accredited for use with verified/EC-type approved balances.
 : For more information on the full range of accessories including printers, software, density control kit for the LA series, external keyboard and PC connection cable, please visit our website or contact your local Avantor sales office.
The balances are also available with a factory calibration certificate. Please contact Avantor for more information about this option.
 : Supplied with mains adapter, universal plug and plastic 'in use' protective cover. Calibration weights are available from Avantor. Please see 'Calibration weights'.
 : Models with a large graphic display are denoted by G in the model name. Similarly i shows internal calibration. FC models are supplied with a factory calibration certificate. See website for details of the full range of models.
  • Model:
    LA 124 *
  • Pan/Platform Size:
    Ø 80 mm
  • Seq:
    01
  • Readability:
    0,1 mg
  • Weighing cap.:
    120 g
  • Linearity:
    0,3 mg
Complementary Products
  •  
    Description
    W×D×H
    Weight
  • 611-2553
    Density kit DT002 (solids and liquids) for VWR balances with readability 0,001; 0,0001 or 0,00001 g (1; 0,1 or 0,01 mg)
    1 * 1 KIT - 959,00 €
  • 611-3446
    ION-15A, ioniser for removing static build-up of weighing samples
    0,31 kg
    310
    1 * 1 items - 866,00 €
Accessories
  •  
    Description
    Print width
    W×D×H
  • 630-1485
    Thermal printer TLP-50 for VWR LA, LP, IS, TA, TS, TP, SM balances and MB/MBT moisture analysers, EU-plug + UK-plug
    54 mm
    106×184×110 mm
    1 * 1 items - 497,00 €
  •  
    Description
  • 611-2795
    Paper roll for DPP-250 printer (611-2793), 1 roll
    1 * 1 items - 15,80 €
  •  
    Description
  • 630-1487
    Labels, self-adhesive, 54×24 mm, 500 on a roll for thermal printer
    1 * 1 items - 23,60 €
  •  
    Description
  • 611-3824
    Complete communication kit with i-Weight software to connect a balance to a computer. Enables data collection, statistics, graphs and exports results to Excel. Functions: Piece counting, Density, Peak hold, Limits set. OS: Windows 7, 8, 10 (32&64). Note not suitable for use with moisture analysers
    1 * 1 items - 259,00 €
  •  
    Description
    For
  • 611-3864
    Ioniser panel, allows user to retrofit an integrated ioniser
    VWR semi-micro, analytical or precision balances fitted with draftshields which have 3 sliding doors
    1 * 1 items - 796,00 €
  •  
    Description
  • 630-2666
    Plastic cover for M,MG analytical
    1 * 1 items - 21,20 €
  •  
    Description
    For
  • 611-3662
    Roll of paper
    TLP-50 printer (630-1485)
    1 * 1 Roll - 18,60 €
  •  
    Description
  • 611-2786
    Serial RS232 to USB converter
    1 * 1 items - 99,40 €