Order Entry
Export
Orders LinkContactUsLinkComponent
WetGrip labels

WetGrip labels

WetGrip printer labels for microscope slides, Paper, Laser, White, 22×22 mm
Catalog # DIVEGRLA-1000
Supplier:   DIVERSIFIED BIOTECH
WetGrip labels
Catalog # DIVEGRLA-1000
WetGrip labels

WetGrip printer labels for microscope slides, Paper, Laser, White, 22×22 mm

Price
184,00 €
Log in for availability
1 * 2.400 items
Product Details & Documents

These WetGrip labels grip to moist, frosty or frozen surfaces.

  • Temperature resistant to −80 °C
  • Compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes

Available in white laser sheets, white direct thermal rolls, or white hand writable rolls.

Specifications
  • Printing method:
    Laser
  • Colour:
    White
  • Description:
    WetGrip printer labels for microscope slides
  • Dimensions:
    22×22 mm
  • Material:
    Paper
  • Seq:
    07