Reagent troughs
Disposable reagent trough, Tecan Standard, 100 ml, Grey
Catalog # TCAN10613049
Supplier: Tecan
Jump to:
Product Details & Documents
These disposable PP troughs are designed to optimise the performance and productivity of Tecan liquid handling platforms.
Specifications
- Capacity:100 ml
- Colour:Grey
- Description:Disposable reagent trough, Tecan Standard
- Seq:03