Spillage trays
Collecting tray with base insert, PEDH-EC material, Black, Electrically conductive, with base insert, 355×445×135 mm, 285×385×95 mm
216-0925
: S.C.A.T.
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
- Electrically conductive available
- Available with base insert; the canisters always stay dry
- Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
- Type:Electrically conductive, with base insert
- W×D×H ext.:355×445×135 mm
- Colour:Black
- Description:Collecting tray with base insert, PEDH-EC material
- Seq:04
- W×D×H int.:285×385×95 mm



