Spillage trays
Collecting tray, PEDH material, Black, Electrically conductive, without base insert, 325×225×215 mm, 300×200×200 mm
Catalog # 216-0924
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
- Electrically conductive available
- Available with base insert; the canisters always stay dry
- Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Type:Electrically conductive, without base insert
- W×D×H ext.:325×225×215 mm
- Colour:Black
- Description:Collecting tray, PEDH material
- Seq:03
- W×D×H int.:300×200×200 mm



