Orders LinkContactUsLinkComponent
Slide boxes, SP Bel-Art
Slide box with removable lid, 50
Catalog # BELAH445760050
Slide boxes, SP Bel-Art
Catalog # BELAH445760050
Slide boxes, SP Bel-Art
Slide boxes, SP Bel-Art
Slide box with removable lid, 50
Price
97,80 €
Log in for availability
1 * 1 items
Product Details & Documents
These slide boxes provide maximum protection for microscope slides.
  • Heavy walls will not bend, break, or crack
  • Unaffected by humidity or insect proof
  • Numbered index allows for easy identification
Specifications
  • No. of slides:
    50
  • Description:
    Slide box with removable lid
  • Seq:
    02