Orders LinkContactUsLinkComponent
1,3-Dibromopropane ≥98%
Glass bottle for solids, 1 kg
  ABCRAB109197.1000
 :   abcr
1,3-Dibromopropane ≥98%
  ABCRAB109197.1000
 :  abcr
 :  109-64-8
1,3-Dibromopropane ≥98%
1,3-Dibromopropane ≥98%
Glass bottle for solids, 1 kg
 :  109-64-8
1 * 1 kg
This product has been blocked by your organization. Please contact your purchasing department for more information
Purity: 98% CAS No.: 109-64-8 Molecular Formula: C3H6Br2 Formula Weight: 201.90 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.99 Refractive Index: 1.5230 Appearance: Colourless Melting Point: -34° Boiling Point: 167° Beilstein Reference: 635662 Merck Reference: 14,9713 UN No.: 1993 MDL No.: MFCD00000255
  •  :109-64-8
  •  :BrH₂CCH₂CH₂Br
  •  :MFCD00000255
  •  :201.89 g/mol
  •  :14,9713
  •  :167 °C
  •  :–34 °C
  •  :1.99
  •  :54 °C (130 °F)
  •  :Ambient
  •  :203-690-3
  •  :1993
  •  :3,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg