Personal eyewash
Eyewash solution, sterile, 500 ml, twin pack
Supplier: ORKLA CARE
Jump to:
Product Details & Documents
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
- Quick and easy to open
- Eye cup protected from dust
- Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
- Neutralises the pH value
Expiry date: 4 years if container not opened.
* Bottle holder is supplied without eyewash.
Documents
Specifications
- Description:Eyewash solution, sterile, 500 ml, twin pack
- Seq:02



