Order Entry
Export
Orders LinkContactUsLinkComponent
 

Silylation reagents

Trimethylsilyl N-trimethylsilylacetamidate, pyridine for GC, BSA:Pyridine, 25 ml, 25 mL
Catalog # HYPETS-49012
Silylation reagents
Catalog # HYPETS-49012
CAS Number:  
Silylation reagents

Trimethylsilyl N-trimethylsilylacetamidate, pyridine for GC, BSA:Pyridine, 25 ml, 25 mL

CAS Number:  
Price
943,00 €
Log in for availability
1 * 25 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    25 mL
  • Capacity:
    25 ml
  • Description:
    Trimethylsilyl N-trimethylsilylacetamidate, pyridine for GC, BSA:Pyridine

Specifications