Order Entry
Export
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, HyperSep™ SCX

Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 1 ml, 50 mg
  HYPE60108-420
Solid phase extraction cartridges, HyperSep™ SCX
  HYPE60108-420
Solid phase extraction cartridges, HyperSep™ SCX
Solid phase extraction cartridges, HyperSep™ SCX

Cartridge, SCX, 40 - 60 µm, 1 ml, 50 mg

 
295,00 €
   
1 * 100 items
 

A strong cation exchange sorbent for extraction of charged basic compounds.

  • Benzene ring provides potential for nonpolar interactions
  • Low pKa value sorbent
 
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    1 ml
  • Seq:
    01
  • Bed weight:
    50 mg
  • Phase:
    SCX