HPLC syringes
Autosampler syringe, -, 500 µl, - mm, AS1000, AS3000
Catalog # 555-8239
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
- Easy and accurate
- Reproducible manual injections
Applications include industrial chromatography.
Specifications
- For:AS1000, AS3000
- Gauge:-
- Volume:500 µl
- Description:Autosampler syringe
- Length:- mm
- Seq:08