Order Entry
Export
Orders LinkContactUsLinkComponent
 

HPLC syringes

Standard fixed needle, 22, 50 µl, 50 mm, 90° blunt end, Rheodyne/Valco™
  555-8443
HPLC syringes
  555-8443
HPLC syringes
HPLC syringes

Standard fixed needle, 22, 50 µl, 50 mm, 90° blunt end, Rheodyne/Valco™

 
80,20 €
   
1 * 1 items
 

These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.

  • Easy and accurate
  • Reproducible manual injections

Applications include industrial chromatography.

 
  • For:
    Rheodyne/Valco™
  • Gauge:
    22
  • Type of tip:
    90° blunt end
  • Volume:
    50 µl
  • Description:
    Standard fixed needle
  • Length:
    50 mm
  • Seq:
    03