Orders LinkContactUsLinkComponent
Inlet base seals
Siltek®-treated Cross inlet base seals, 0,8 mm ID
Catalog # 555-9219
Inlet base seals
Catalog # 555-9219
Inlet base seals
Inlet base seals
Siltek®-treated Cross inlet base seals, 0,8 mm ID
Price
480,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.
Specifications
  • Description:
    Siltek®-treated Cross inlet base seals, 0,8 mm ID
  • Seq:
    10