Orders LinkContactUsLinkComponent
Waste caps
Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter, S 61, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter, PP
Catalog # JOURJR-S-16113
Supplier:   VICI JOUR RESEARCH
Waste caps
Catalog # JOURJR-S-16113
Supplier:  VICI JOUR RESEARCH
Waste caps
Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter, S 61, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter, PP
Price
240,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
 : Barbed adapters must be ordered separately.
Specifications
  • Thread:
    S 61
  • Description:
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
  • Material:
    PP
  • Seq:
    13
  • Connector:
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter