Orders LinkContactUsLinkComponent
Waste caps
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, B 83, 4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Catalog # JOURJR-S-18304
Supplier:   VICI JOUR RESEARCH
Waste caps
Catalog # JOURJR-S-18304
Supplier:  VICI JOUR RESEARCH
Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, B 83, 4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Price
575,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Specifications
  • Thread:
    B 83
  • Description:
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
  • Material:
    PP
  • Seq:
    03
  • Connector:
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing