Waste caps
Waste cap, includes O-ring, 1/4" nuts, ferrules and plug, B 53, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing, PP
Catalog # JOURJR-S-15303
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Specifications
- Thread:B 53
- Description:Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
- Material:PP
- Seq:01
- Connector:3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing