Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thermal transfer printer labels, type B-423

Printer labels, B-423, THT-55-423-1, Polyester, Permanent acrylic, Thermal transfer, White, 360X-Plus II, 101×50 mm, 1000 labels/roll
Catalog # BRDY030245
Supplier:   Brady
Thermal transfer printer labels, type B-423
Catalog # BRDY030245
Supplier:  Brady
Thermal transfer printer labels, type B-423
Thermal transfer printer labels, type B-423

Printer labels, B-423, THT-55-423-1, Polyester, Permanent acrylic, Thermal transfer, White, 360X-Plus II, 101×50 mm, 1000 labels/roll

Price
252,00 €
Log in for availability
1 * 1 Roll
Product Details & Documents

Polyester labels for thermal transfer printers.

  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon separate, not in the same enclosure as the tape/sleeves

This material is UL, CSA, and AGA approved with the respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.

Specifications
  • Printer model:
    360X-Plus II
  • Printing method:
    Thermal transfer
  • Type:
    B-423
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    101×50 mm
  • Material:
    Polyester
  • Seq:
    22
  • Model:
    THT-55-423-1
  • Packed:
    1000 labels/roll
  • Adhesive:
    Permanent acrylic