Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
250 ml, 22 mm, 45×60×125 mm
Catalog # SEMA2024
Supplier: SEMADENI
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Catalog # SEMA2024
Supplier: SEMADENI
Jump to:
Product Details & Documents
HDPE, translucent, with thread.
- Good chemical resistance
- Good temperature resistance
Ordering information: Screw caps must be ordered separately.
Specifications
- Capacity:250 ml
- W×D×H:45×60×125 mm
- Neck I-Ø:22 mm



