drum gripper for forkliftfor easy and safe transport of steel barrels, jaw capacity: 420 - 610 mm load: 350 kg weight: 6,0 kg 1 * 1 items
Catalog # DUEH1.31.103
drum gripper for forkliftfor easy and safe transport of steel barrels, jaw capacity: 420 - 610 mm load: 350 kg weight: 6,0 kg 1 * 1 items
Catalog # DUEH1.31.103