ContactUsLinkComponent
safety cabinet with folding doors outer carcass: 1195 x 595 x 2080 (wxdxh) mm, inner carcass: 1099 x 446 x 1830 (wxdxh) mm, outer ca rcass, sheet steel, powder-coated in ral 7035 light grey, folding doors in ral 1018 zinc yellow, inner carcass ral 70 1 * 1 items
  DUEH29-201262-086
 :  DUPERTHAL
safety cabinet with folding doors outer carcass: 1195 x 595 x 2080 (wxdxh) mm, inner carcass: 1099 x 446 x 1830 (wxdxh) mm, outer ca rcass, sheet steel, powder-coated in ral 7035 light grey, folding doors in ral 1018 zinc yellow, inner carcass ral 70 1 * 1 items
  DUEH29-201262-086
 :  DUPERTHAL