ContactUsLinkComponent
safety cabinet with wing door. outer carcass: 1195 x 595 x 2080 (wxdxh) mm, inner carcass: 1099 x 446 x 1830 (wxdxh) mm, outer carca ss, sheet steel, powder-coated in ral 7035 light grey, wing doors in ral 1018 zinc yellow, inner carcass in ral 7035 1 * 1 items
Catalog # DUEH29-201260-068
Supplier:  DUPERTHAL
undefined
safety cabinet with wing door. outer carcass: 1195 x 595 x 2080 (wxdxh) mm, inner carcass: 1099 x 446 x 1830 (wxdxh) mm, outer carca ss, sheet steel, powder-coated in ral 7035 light grey, wing doors in ral 1018 zinc yellow, inner carcass in ral 7035 1 * 1 items
Catalog # DUEH29-201260-068
Supplier:  DUPERTHAL
Related Information