Order Entry
Export
Orders LinkContactUsLinkComponent
DirectPCR® DNA extraction system

DirectPCR® DNA extraction system

DirectPCR®-Yolk Sac, optimised for lysis of mouse embryonic material, 100 ml, 100 mL
Catalog # 732-3258
Supplier:   VIAGEN BIOTECH
DirectPCR® DNA extraction system
Catalog # 732-3258
Supplier:  VIAGEN BIOTECH
CAS Number:  
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system

DirectPCR®-Yolk Sac, optimised for lysis of mouse embryonic material, 100 ml, 100 mL

CAS Number:  
Price
336,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

DirectPCR® DNA Lysis Reagents offer a quick and cost-effective 'single-tube' system for the rapid preparation of PCR template DNA from mouse tails, ears, embryonic material or mammalian cells.

  • 'Single-tube' system for reduced number of pipetting steps
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions or time-consuming alcohol precipitations
  • Reliable and absolutely reproducible procedure
  • High yields
  • Lysate can directly be used for genotyping PCRs

NOTE: 1 µl lysate is sufficient for subsequent genotyping PCR reactions.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    DirectPCR®-Yolk Sac, optimised for lysis of mouse embryonic material, 100 ml

Specifications