Order Entry
Export
Orders LinkContactUsLinkComponent
 

DirectPCR® DNA extraction system

DirectPCR®-Ear, optimised for lysis of mouse ear tissue, 50 ml, 50 mL
  732-2959
DirectPCR® DNA extraction system
  732-2959
 :  
DirectPCR® DNA extraction system
DirectPCR® DNA extraction system

DirectPCR®-Ear, optimised for lysis of mouse ear tissue, 50 ml, 50 mL

 :  
 
336,00 €
   
1 * 50 mL
 

DirectPCR® DNA Lysis Reagents offer a quick and cost-effective 'single-tube' system for the rapid preparation of PCR template DNA from mouse tails, ears, embryonic material or mammalian cells.

  • 'Single-tube' system for reduced number of pipetting steps
  • Suitable for the simultaneous processing of multiple samples
  • No need for organic extractions or time-consuming alcohol precipitations
  • Reliable and absolutely reproducible procedure
  • High yields
  • Lysate can directly be used for genotyping PCRs

NOTE: 1 µl lysate is sufficient for subsequent genotyping PCR reactions.

 

 

  • Pk:
    50 mL
  • Description:
    DirectPCR®-Ear, optimised for lysis of mouse ear tissue, 50 ml