ContactUsLinkComponent
Shodex IEC DEAE-5025 - Ion exchange preparative HPLC column, polymer based - ID: 50mm length: 300mm - Functional Group: Diethylamino ethyl - Base Material: Polyhydroxymethacrylate - Particle Size (µm): 37 - Pore Size (A): - 1 * 1 items
Catalog # SHOWF6548051
Supplier:  Resonac
Shodex IEC DEAE-5025 - Ion exchange preparative HPLC column, polymer based - ID: 50mm length: 300mm - Functional Group: Diethylamino ethyl - Base Material: Polyhydroxymethacrylate - Particle Size (µm): 37 - Pore Size (A): - 1 * 1 items
Catalog # SHOWF6548051
Supplier:  Resonac