Order Entry
Export
Orders LinkContactUsLinkComponent
DNA prep for PCR - tissue, Extracta™

DNA prep for PCR - tissue, Extracta™

Extracta™ DNA prep for PCR - tissue, 25 mL
Catalog # 733-2160
Supplier:   Quantabio
DNA prep for PCR - tissue, Extracta™
Catalog # 733-2160
Supplier:  Quantabio
CAS Number:  
DNA prep for PCR - tissue, Extracta™

Extracta™ DNA prep for PCR - tissue, 25 mL

CAS Number:  
Price
158,00 €
Log in for availability
1 * 25 mL
Product Details & Documents

The Extracta™ DNA prep for PCR - tissue consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva.

  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, high resolution melt analysis, endpoint PCR and other applications
  • Works with wide range of tissue
  • Optional 10 minute protocol for high throughput applications

The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.

Documents

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Extracta™ DNA prep for PCR - tissue
  • Cat. No.:
    733-2160
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Extraction for direct PCR
  • Description:
    Extracta™ DNA prep for PCR - tissue
  • Sample type:
    Cultured cells|Human/animal cells and tissues

Specifications