Orders LinkContactUsLinkComponent
Pipette tips station, Corning® DeckWorks™
Pipette tips station, Corning® DeckWorks™
Round tip, 1 - 200 µl, -, Transparent, 960 tips/pack
Catalog # CORN4142
   Waste Reducing
Supplier:   Corning
Pipette tips station, Corning® DeckWorks™
Catalog # CORN4142
Supplier:  Corning
   Waste Reducing
Pipette tips station, Corning® DeckWorks™
Pipette tips station, Corning® DeckWorks™
Round tip, 1 - 200 µl, -, Transparent, 960 tips/pack
Price
381,00 €
Log in for availability
1 * 4.800 items
Product Details & Documents
These tip stations maximise bench space while minimising plastic waste. The stations contain 960 tips, available in extended length 10 µl and traditional 200 µl sizes.
  • Ideal single and multi-channel pipetting stations
  • Five-layer deck design (2×96) matches the microtiter plate format
  • Stable pipetting platform fits easily into sterile hood environments
  • Lids can be snap-positioned to expose a single side (96 tips) only
Applications include pre- and post-PCR procedures, cell culture and assays requiring large tip quantities. These tip stations are offered both sterile and non sterile.
Certified Free of: RNase, DNase, Pyrogens, PCR Inhibitors.
Both the hinged racks and reload decks within this system are manufactured with recycled plastic. The entire DeckWorks system is comprised of seven sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
Ordering information: Corning DeckWorks pipette tips are also available in bulk, hinged rack, barrier hinged rack, reload and reload starter kit configurations.
Documents
Specifications
  • Packed:
    960 tips/pack
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Seq:
    03
  • Tip style:
    Round tip
  • Tip colour:
    Transparent
  • Cat. No.:
    CORN4142
  • Environmentally Preferable:
    Waste Reducing
Accessories
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Pipette tip racks, empty, for 10/200/300 µl tips
    Pipette tip racks, empty, for 10/200/300 µl tips
    CORN4145
    Pipette tips station, DeckWorks™
    Log in for availability
    1 * 24 items - 87,00 €
Product Information
  • Catalog #
    For use with
    Availability
    Price
  • Pipette tip racks, empty, with MultiGuard, for 1000 µl tips
    Pipette tip racks, empty, with MultiGuard, for 1000 µl tips
    CORN4146
    Pipette tips station, DeckWorks™
    Log in for availability
    1 * 16 items - 79,70 €