Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
PBS buffer solution (phosphate buffered saline) pH 7,4, 50
Catalog # SIALP3813-5X10PAK
Supplier: Merck
Buffer, PBS solution (phosphate buffered saline) pH 7,4, Sigma-Aldrich®
Catalog # SIALP3813-5X10PAK
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:50
- Description:PBS buffer solution (phosphate buffered saline) pH 7,4



