Order Entry
Export
Orders LinkContactUsLinkComponent
NMR STD/0.1% ETHYLBENZENE/CDCL3 100% & 1 * 1 Each

NMR STD/0.1% ETHYLBENZENE/CDCL3 100% & 1 * 1 Each

Catalog # SIAL708054-8IN
Supplier:   Merck
NMR STD/0.1% ETHYLBENZENE/CDCL3 100% & 1 * 1 Each
Catalog # SIAL708054-8IN
Supplier:  Merck
CAS Number:  
NMR STD/0.1% ETHYLBENZENE/CDCL3 100% & 1 * 1 Each
CAS Number:  
Price
371,00 €
Log in for availability
1 * 1 Each
Product Details & Documents