Order Entry
Export
Orders LinkContactUsLinkComponent
Connectors

Connectors

Connector, 25 mm TC to 6,4 mm hose barb, Polysulfone
Catalog # KRB4-2PS
Supplier:   Cytiva
Connectors
Catalog # KRB4-2PS
Supplier:  Cytiva
Connectors

Connector, 25 mm TC to 6,4 mm hose barb, Polysulfone

Price
152,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Connector, 25 mm TC to 6,4 mm hose barb
  • Material:
    Polysulfone
  • Seq:
    01