Order Entry
Export
Orders LinkContactUsLinkComponent
Closures for snap ring vials, Supelco®

Closures for snap ring vials, Supelco®

PP sealed cap, 11 mm Snap ring vials, PTFE/silicone, 11,5 mm
Catalog # SIAL24758
Supplier:   Merck
Closures for snap ring vials, Supelco®
Catalog # SIAL24758
Supplier:  Merck
Closures for snap ring vials, Supelco®

PP sealed cap, 11 mm Snap ring vials, PTFE/silicone, 11,5 mm

Price
40,80 €
Log in for availability
1 * 1 Each
Product Details & Documents
Specifications
  • Ø:
    11,5 mm
  • For:
    11 mm Snap ring vials
  • Septum:
    PTFE/silicone
  • Type:
    PP sealed cap
  • Seq:
    05